Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

330602

Avanti

OxPC ELISA (Black)

Avanti Research - A Croda Brand

Synonym(s):

OxPC ELISA, OxPAPC ELISA Snoopers, Black, for use in fluorescence applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

form

liquid

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 1 × (ELISA strip packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

ELISA Snoopers are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid binding proteins. This format provides the following advantages:
  • quantitative lipid-protein binding studies
  • assemble a 96-well plate using only the lipids of interest for binding studies
  • flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.

The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.

Application

OxPC ELISA (Black) has been used in competitive binding assay for quantification of E06 binding lipids from rat paw samples.

Packaging

ELISA Strip Packaged in Foil Pouch (330602-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antinociception by the anti-oxidized phospholipid antibody E06
Mohammadi M, et al.
British Journal of Pharmacology, 175(14), 2940-2955 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service