Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z765309

Tygon® formula E-3603 laboratory tubing

I.D. × O.D. × wall thickness 1/4 in. × 5/8 in. × 3/16 in.

Synonym(s):

DEHP tubing alternative, chemical resistant tubing, laboratory tubing, vacuum tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105108
NACRES:
NB.35

packaging

pkg of 50 feet

manufacturer/tradename

Tygon

I.D. × O.D. × wall thickness

1/4 in. × 5/8 in. × 3/16 in.

Looking for similar products? Visit Product Comparison Guide

General description

TYGON® E-3603 non-DEHP laboratory and vacuum tubing delivers the same superior performance you′ve come to expect from TYGON products - but now in a formulation that contains a widely accepted substitute for DEHP plasticizer.

TYGON® E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion, handles virtually all inorganic chemicals found in the lab, and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.

TYGON® E-3603 vacuum tubing shares all these performance features and has extra heavy walls that will withstand a full vacuum at room temperature.

Features/Benefits:
  • Alternative to DEHP tubing
  • New formulation delivers the same excellent performance
  • Contains no BPA
  • Outstanding chemical resistance
  • Lot-to-lot consistency for reproducible results
  • Increases productivity in peristaltic pumps - outlasts other clear tubing 2 to 1
  • Smooth, polished inner bore
  • Non-oxidizing and non-contaminating
  • Slips easily over fittings and grips securely
  • Coils marked at 1-foot intervals for easy measuring
  • Meets FDA criteria

Typical Applications:
  • General laboratory
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain lines

Legal Information

Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service