Key Documents
Z742413
High pressure adapter for Q-Tube
for 12 mL Q-Tube, max. pressure 180 psi
About This Item
UNSPSC Code:
51111800
NACRES:
NB.51
material
aluminum
manufacturer/tradename
Q Labtech 100-TA0180-120
parameter
-200 psi pressure (max)
180 psi max. pressure
General description
Pressure Adapter-12 mL-180 psi is designed to fit on 12 mL systems. This blue anodized aluminum adapter can hold up to 180 psi internal pressure. This higher threshold is ideal for hydrogenation and reactions involving ammonia hydroxide, methylamine, dimethylamine, formaldehyde, acetaldehyde, and hydrogen chloride.
Legal Information
Q-Tube is a trademark of Q Labtech LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)