Documents
Z556378
Sigma-Aldrich
Aldrich® Sure/Stor adapter
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Joint
joint: ST/NS24/40
description
with PTFE J Young valve and SafetyBarb®
hose connector
Related Categories
General description
Converts round-bottom flasks to Sure/Stor flasks.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Stor is a trademark of Sigma-Aldrich Co. LLC