Key Documents
Z150703
Sigma-Aldrich
Window for PerkinElmer® and Wilks holders
KBr, Undrilled
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
material
KBr
rectangular
description
Polished
technique(s)
IR spectroscopy: suitable
L × W × H
38.5 mm × 19.5 mm × 4 mm
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)