Key Documents
Z104329
Sigma-Aldrich
Aldrich® reaction vessel bubbler
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/40
capacity
5-7 mL
General description
Excellent for closing off a reaction vessel from the atmosphere.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)