Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

WHA2812259

Whatman® high-purity glass microfiber extraction thimbles

silica thimbles, tapered, 25 x 90 mm, 10/pk

Synonym(s):

Z742074, extraction thimbles, glass fiber filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

material

quartz

sterility

non-sterile

packaging

pack of 10

manufacturer/tradename

Whatman 2812-259
Whatman Article No. 28419019 (US reference)

O.D. × H

25 mm × 90 mm

Looking for similar products? Visit Product Comparison Guide

General description

High-purity Whatman quartz thimbles from Cytiva support accurate, reproducible emission testing by providing low extractable levels and temperature resistance up to 800°C.

Quartz thimble filter size

Each Whatman quartz thimble from Cytiva is made from high-purity quartz with low heavy metal concentrations. Thimbles are 90 mm in length, tapered for stack sample loading, and have an outside diameter of 25 mm.

Application

  • Smoke stack gas monitoring
  • Soxhlet extraction
  • Analyzing pesticide residues
  • Determining oil/fat content of foods (e.g. French fries)
  • Analysis of oil and grease in solid wastes

Features and Benefits

  • Appropriate for manual sampling in aggressive environments
  • Suitable for both solvent extraction and air sampling applications
  • High-purity quartz/silica, for high-temperature resistance (up to 800°C)
  • No binders or chemical additives
  • Low heavy metal content
  • High collection efficiency

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service