Key Documents
TEM-FCF100NI50
Sigma-Aldrich
Formvar/Carbon Supported Nickel Grids
size 100 mesh, box of 50 ×
Synonym(s):
Formvar/Carbon 100 Mesh, Ni, 50pk
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Pricing and availability is not currently available.
packaging
box of 50 ×
size
100 mesh
General description
A formvar coated TEM grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Thickness: 10nm Formvar and 1nm Carbon
  • Choose from one of the most recent versions:
    Certificates of Analysis (COA)