Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF04687858

Chromium

powder, max. particle size 45 micron, weight 200 g, min. particle size 38 micron

Synonym(s):

CR006025

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$449.00

$449.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$449.00

About This Item

Empirical Formula (Hill Notation):
Cr
CAS Number:
Molecular Weight:
52.00
MDL number:
UNSPSC Code:
12141709
PubChem Substance ID:
NACRES:
NA.23

$449.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

form

powder

manufacturer/tradename

Goodfellow 046-878-58

resistivity

12.7 μΩ-cm, 20°C

weight

200 g

max. part. size

45 μm

bp

2672 °C (lit.)

mp

1857 °C (lit.)

density

7.14 g/mL at 25 °C (lit.)

SMILES string

[Cr]

InChI

1S/Cr

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service