Key Documents
DWK213911772
DURAN® measuring cylinder ungraduated with hexagonal base
glass, cylinder capacity 50 nL, unstamped
Synonym(s):
213911772
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
glass
feature
lid: no
packaging
case of 10 ea
manufacturer/tradename
DURAN® 213911772
cylinder capacity
50 nL
General description
Calibration is based on the poured in volume (“In”) at a + 20 °C reference temperature. A uniform wall thickness over the entire measuring range prevents wedge errors. Fired-on, highly durable white print. Easy-to-read scale with graduation marks to estimate the content. The volume content tolerances conform to accuracy Class B.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)