Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CPC-606

Sigma-Aldrich

Natural PC Liposomes

Phosphatidylcholine (PC)-based - Brain PC:Cholesterol (70:30 molar ratio 5mM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

biological source

Porcine brain

Quality Level

composition

Phosphate buffered saline

concentration

5 mM

impurities

30 mol % Cholesterol
70 mol % L-alpha-phosphatidylcholine (PC)

particle size

100 nm

pH

7.4

Looking for similar products? Visit Product Comparison Guide

General description

Liposomes are extensively used to study the interaction of proteins, peptides and other molecules with the surface of a lipid membrane.

The natural PC liposome catalog are made from phosphatidylcholine (PC) lipids extracted from egg yolk, soybean, tissues and organs such as brain, heart and liver, with and without cholesterol. Adding 30% cholesterold makes the liposomes serum stable and is necessary when the liposomes are used in the presence of serum.

Application

Drug delivery
Lipid-protein interactions

Storage and Stability

Liposomes should never be frozen. Liposomes should be stored in the dark at 4°C, except when brought to room temperature for brief periods prior to use.

Liposomes are made under sterile conditions. If you need to take multiple aliquots out of the vial, it is advised to take extreme care in not contaminating the vial. It is recommended to handle the vial under a sterile hood to maintain the sterility of the product. Liposomes should never be frozen. Ice crystals that form during freezing will rupture the lipid membrane of the liposomes and change the size of liposomes particles.

Legal Information

Cellsome is a trademark of Encapsula NanoSciences
Product of Encapsula Nanosciences

Disclaimer

For research use only

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service