Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

BMSBSH300E

Sigma-Aldrich

MyBlock Mini dry bath with cooling

AC/DC input 100 V / 240 V AC, Schuko plug

Synonym(s):

block heater/cooler, mini block heater/cooler, personal dry bath

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NB.76

manufacturer/tradename

Benchmark Scientific BSH300-E

parameter

-25 °C temp. range (100 °C)

Looking for similar products? Visit Product Comparison Guide

General description

  • Capable of heating as well as cooling test samples
  • Minimum Programmable temp.: 0 C
  • Compact, fits in the palm of your hand
  • Exchangeable blocks for 0.2 to 50ml tubes

Legal Information

MyBlock is a trademark of Benchmark Scientific, Inc.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service