MilliporeSigma
All Photos(4)
Key Documents
BAH369460010
SP Bel-Art® Sterileware Long Handled Sampling Spoon
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm), sterile, pack of 10 ea (individually wrapped)
Synonym(s):
Lab Scoop, dispensing tool, spoon
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
white polystyrene
sterility
sterile
packaging
pack of 10 ea (individually wrapped)
manufacturer/tradename
Bel-Art H36946-0010
L × W
17.8 cm (7 in.) × 1.5625 in. (40 mm)
nominal capacity
14.79 mL , 3tsp
General description
Designed to gather small amounts of sample material, especially in hard to reach containers.
Features and Benefits
  • Long handle reaches into jars, bottles and other confined areas
  • Minimizes the potential of personal contact with sampled material
  • Sterile, FDA grade polystyrene safe for food, drug and cosmetic use
  • Individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing
Note: Sterilized products are not returnable.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP