Key Documents
BAF131641419
Scienceware® biohazard disposal bags
width × H × thickness 14 in. (360 mm) × 19 in. (480 mm) × 0.04 mm (1.5 mil), pack of 200 ea, translucent red polypropylene
Synonym(s):
Scienceware® biohazard disposal bags
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
translucent red polypropylene
feature
label (With Biohazard Warning Label and Sterilization Indicator Patch)
Fits waste can 2.0 gallon
packaging
pack of 200 ea
manufacturer/tradename
Bel-Art F13164-1419
capacity
2.0 gal
7.6-15.1 L (2-4 gal)
width × H × thickness
14 in. (360 mm) × 19 in. (480 mm) × 0.04 mm (1.5 mil)
General description
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
Features and Benefits
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • High temperature steam autoclavable to 135°C (275°F)
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • Made in USA
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)