Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900765

Sigma-Aldrich

4,8-Bis(3,5-dioctyl-2-thienyl)-2,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b′]dithiophene

Synonym(s):

2,2′-(4,8-Bis(3,5-dioctylthiophen-2-yl)benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H96B2O4S4
CAS Number:
Molecular Weight:
1055.31
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

>98%

form

solid

storage temp.

2-8°C

SMILES string

CC(C(C)(C)O1)(C)OB1C(S2)=CC3=C2C(C4=C(CCCCCCCC)C=C(CCCCCCCC)S4)=C(C=C(B5OC(C)(C)C(C)(C)O5)S6)C6=C3C7=C(CCCCCCCC)C=C(CCCCCCCC)S7

InChI

1S/C62H96B2O4S4/c1-13-17-21-25-29-33-37-45-41-47(39-35-31-27-23-19-15-3)69-55(45)53-49-43-51(63-65-59(5,6)60(7,8)66-63)72-58(49)54(50-44-52(71-57(50)53)64-67-61(9,10)62(11,12)68-64)56-46(38-34-30-26-22-18-14-2)42-48(70-56)40-36-32-28-24-20-16-4/h41-44H,13-40H2,1-12H3

InChI key

OYNKXAWHHLJVGD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A polybenzo[1,2-b:4,5-b']dithiophene derivative with deep HOMO level and its application in high-performance polymer solar cells.
Lijun Huo et al.
Angewandte Chemie (International ed. in English), 49(8), 1500-1503 (2010-01-27)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service