Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

756261

Sigma-Aldrich

3,5-Dihydroxycyclohexanecarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

135-140 °C

functional group

carboxylic acid
hydroxyl

SMILES string

OC1CC(O)CC(C1)C(O)=O

InChI

1S/C7H12O4/c8-5-1-4(7(10)11)2-6(9)3-5/h4-6,8-9H,1-3H2,(H,10,11)

InChI key

JOIXGDNGAXHWPO-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service