Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

754471

Sigma-Aldrich

Potassium 4-(2-methoxyethylcarbamoyl)phenyltrifluoroborate

97%

Synonym(s):

Potassium 4-(2-methoxyethylamine-1-carbonyl)phenyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12BF3KNO2
Molecular Weight:
285.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

206-211 °C

storage temp.

2-8°C

SMILES string

[K+].COCCNC(=O)c1ccc(cc1)[B-](F)(F)F

InChI

1S/C10H12BF3NO2.K/c1-17-7-6-15-10(16)8-2-4-9(5-3-8)11(12,13)14;/h2-5H,6-7H2,1H3,(H,15,16);/q-1;+1

InChI key

HOIRNIUHFHDIIW-UHFFFAOYSA-N

Application

Potassium trifluoroborate salt is commonly used in palladium catalyzed carbon-carbon formation (Suzuki Reaction) and display longer shelf stability than their corresponding boronic acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gary A Molander et al.
Accounts of chemical research, 40(4), 275-286 (2007-01-30)
Organotrifluoroborates represent an alternative to boronic acids, boronate esters, and organoboranes for use in Suzuki-Miyaura and other transition-metal-catalyzed cross-coupling reactions. The trifluoroborate moiety is stable toward numerous reagents that are often problematic for other boron species. Consequently, remote functional groups

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service