Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

615285

Sigma-Aldrich

1-Octanol-d18

98 atom % D

Synonym(s):

Capryl alcohol-d18

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)7OD
CAS Number:
Molecular Weight:
148.34
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

bp

196 °C (lit.)

mp

-15 °C (lit.)

density

0.940 g/mL at 25 °C

mass shift

M+18

SMILES string

[2H]OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D

InChI key

KBPLFHHGFOOTCA-PBYHGZINSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

176.0 °F

Flash Point(C)

80 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service