Skip to Content
MilliporeSigma
All Photos(1)

Documents

605786

Sigma-Aldrich

Palmitic acid-2,4,6,8,10,12,14,16-13C8

99 atom % 13C

Synonym(s):

Hexadecanoic acid-2,4,6,8,10,12,14,16-13C8

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(CH213CH2)7CO2H
Molecular Weight:
264.37
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

mp

61-64 °C (lit.)

mass shift

M+8

SMILES string

[13CH3]C[13CH2]C[13CH2]C[13CH2]C[13CH2]C[13CH2]C[13CH2]C[13CH2]C(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1+1,3+1,5+1,7+1,9+1,11+1,13+1,15+1

InChI key

IPCSVZSSVZVIGE-ZXWYYLDKSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service