Skip to Content
MilliporeSigma
All Photos(1)

Documents

445894

Sigma-Aldrich

Poly(ethylene glycol) dimethyl ether

average MN 1,000, cross-linking reagent click chemistry

Synonym(s):

Polyethylene glycol, Dimethyl polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol) dimethyl ether, average Mn ~1,000

form

solid

Quality Level

mol wt

average Mn ~1,000

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

refractive index

n20/D 1.46

bp

>250 °C

transition temp

Tm (DSC) 42 °C (at peak)

density

1.08 g/mL at 25 °C

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

CO.OCCO

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

312.8 °F - closed cup

Flash Point(C)

156.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service