Skip to Content
MilliporeSigma
All Photos(1)

Documents

360090

Sigma-Aldrich

2-Methoxy-6-methylaniline

98%

Synonym(s):

6-Methyl-o-anisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(CH3)NH2
CAS Number:
Molecular Weight:
137.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

62 °C/0.15 mmHg (lit.)

mp

26-29 °C (lit.)

SMILES string

COc1cccc(C)c1N

InChI

1S/C8H11NO/c1-6-4-3-5-7(10-2)8(6)9/h3-5H,9H2,1-2H3

InChI key

HKOJYPPTIPJZAZ-UHFFFAOYSA-N

Application

2-Methoxy-6-methylaniline may be used in the synthesis of 7-methoxy-1H-indazole, an inhibitor of nitric oxide synthase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Sopková-de Oliveira Santos et al.
Acta crystallographica. Section C, Crystal structure communications, 58(Pt 11), o688-o690 (2002-11-05)
The crystal structure of 7-methoxy-1H-indazole, C(8)H(8)N(2)O, an inhibitor of nitric oxide synthase, shows that the methoxy group lies in the plane of the indazole system with its methyl group located trans to the indazole N-H group. The crystal packing consists
Selective Oxidation of Aromatic Amines to Nitro Derivatives using Potassium Iodide-tert-Butyl Hydroperoxide Catalytic System.
Reddy KR, et al.
Advanced Synthesis & Catalysis, 351(1-2), 93-96 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service