Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

139408

Sigma-Aldrich

3,5-Dichlorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CHO
CAS Number:
Molecular Weight:
175.01
Beilstein:
2040553
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

63.5-65.5 °C (lit.)

functional group

aldehyde
chloro

SMILES string

[H]C(=O)c1cc(Cl)cc(Cl)c1

InChI

1S/C7H4Cl2O/c8-6-1-5(4-10)2-7(9)3-6/h1-4H

InChI key

CASRSOJWLARCRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of 4-hydroxyl substituent on the singlet chemiexcitation yield in base-catalyzed air oxidation of 3, 5-dichlorophenylpyruvic acids.
Kamiya I, et al.
Bulletin of the Chemical Society of Japan, 57(7), 1740-1744 (1984)
An efficient new enzymatic method for the preparation of ?-aryl-?-amino acid enantiomers.
Tasnadi G, et al.
Tetrahedron Asymmetry, 19(17), 2072-2077 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service