跳转至内容
Merck
BMP-SMAD1/5 Signaling Regulates Retinal Vascular Development.
Biomolecules (2020-03-27)
Andreas Benn, Florian Alonso, Jo Mangelschots, Elisabeth Génot, Marleen Lox, An Zwijsen
摘要
Vascular development is an orchestrated process of vessel formation from pre-existing vessels via sprouting and intussusceptive angiogenesis as well as vascular remodeling to generate the mature vasculature. Bone morphogenetic protein (BMP) signaling via intracellular SMAD1 and SMAD5 effectors regulates sprouting angiogenesis in the early mouse embryo, but its role in other processes of vascular development and in other vascular beds remains incompletely understood. Here, we investigate the function of SMAD1/5 during early postnatal retinal vascular development using inducible, endothelium-specific deletion of Smad1 and Smad5. We observe the formation of arterial-venous malformations in areas with high blood flow, and fewer and less functional tip cells at the angiogenic front. The vascular plexus region is remarkably hyperdense and this is associated with reduced vessel regression and aberrant vascular loop formation. Taken together, our results highlight important functions of SMAD1/5 during vessel formation and remodeling in the early postnatal retina.
材料
货号
品牌
产品描述
Sigma-Aldrich
抗-肌动蛋白, α-平滑肌- Cy3抗体,小鼠单克隆, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-EphB4 antibody produced in goat, affinity isolated antibody, lyophilized powder