主要文件
G006070
GC Analysis of Kennewick Peppermint Essential Oil on SLB®-IL60
suitable for GC
登录查看公司和协议定价
About This Item
技术
gas chromatography (GC): suitable
试验参数
column: SLB-IL60, 30 m x 0.25 mm I.D., 0.25 μm (29505-U)
oven: 75 °C (4 min), then 4 °C/min to 200 °C (10 min)
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: helium, 25cm/ sec set at 110°C
injection: 0.1 μL 100:1 split
liner: 4 mm I.D. cup injector
sample: Kennewick Peppermint Oil, (A.M. Todd Sample), neat
应用
flavors and fragrances
法律信息
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany
分析色谱柱
历史批次信息供参考:
分析证书(COA)