Merck
59269
SSI 高压在线过滤器
10-32 Valco compatible thread
别名:
在线过滤器,CPI 接头,0.5μm,件
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115700
包裝
pkg of 1 ea
接頭
10-32 Valco compatible thread
正在寻找类似产品? 访问 产品对比指南
一般說明
柱前安装。316 不锈钢筛板(0.5μm 孔)无需取下色谱柱柱头即可轻松更换。最大使用压力:15,000psi(1054kg/cm²)。用于 1/16 英寸管。
柱前安装。316 不锈钢筛板(0.5μm 孔)无需取下色谱柱柱头即可轻松更换。用于 1/16 英寸管。最大使用压力:15,000psi(1,054kg/cm2)。
法律資訊
SSI is a trademark of Scientific Systems, Inc.
从最新的版本中选择一种:
分析证书(COA)
Lot/Batch Number
抱歉,我们目前尚未线上提供该产品的COA。
访问文档库
商品
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.
查看所有结果