2635825
1 of 10
Supelco
2637505
进样口衬管,直接 (SPME) 型,直型设计(未包装)
2637501
2633901
进样口衬管,固相微萃取/不分流,用于带 SPL-17 进样器的 17A
2633905
2051305
进样口衬管,不分流
CLS2975243
Corning® 盖玻片
57304
聚丙烯酸酯 (PA) 固相微萃取萃取头
57328-U
SPME 纤维组件二乙烯基苯/羧基/聚二甲基硅氧烷 (DVB/CAR/PDMS)
503258
Supeltex M-2A Capillary Ferrule, General Purpose
57330-U
SPME 光纤支架
SPME/Splitless
pkg of 25 ea
74 mm × 6.35 mm × 0.75 mm
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.