主要文件
Z732443
IKA ® ULTRA-TURRAX ® 分散工具,不锈钢
dispersing for T-25 digital disperser, 10 mm, 1/cs
别名:
disperser blades, tissue grinders, tissue homogenizers
登录查看公司和协议定价
About This Item
UNSPSC代码:
41103800
NACRES:
NB.22
物料
stainless steel
特点
stainless steel rotor/stator
包装
pack of 1
制造商/商品名称
IKA 0000594000
直径
10 mm
正在寻找类似产品? 访问 产品对比指南
一般描述
IKA®S 25 N - 10 G Dispersing tool for use with T 25 digital and easy cleabn ULTRA-TURRAX® dispersing instruments.
特点和优势
  • suitable for use with solvents
  • sterilization methods: all methods
法律信息
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG
历史批次信息供参考:
分析证书(COA)