Merck
RA4384-EP
Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges
A-4-38, aerosol-tight, with 4 x 100 mL round buckets
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103909
特點
to hold 1.5 - 100 mL tubes
to hold x blood collection and cell culture tubes
aerosol-tight
swing bucket rotor
with 4 x 100 mL round buckets
包裝
pack of 1 ea
製造商/商標名
Eppendorf® 022639048 (NA)
Eppendorf® 5702720003 (EU)
參數
3,000 × g max. RCF
技術
centrifugation: suitable
相容性
for use with centrifuge models 5702/5702R/502RH
正在寻找类似产品? 访问 产品对比指南
相关类别
法律資訊
Eppendorf is a registered trademark of Eppendorf AG