主要文件
OH30073974
Ohaus® Pen pH Meter
Model ST10C-C
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115603
包裝
pack of 1 ea
製造商/商標名
Ohaus® 30073974
電導率測量範圍
0.00-19.99 mS/cm
電池電量
4 × 1.5 V, 200 hr life
正在寻找类似产品? 访问 产品对比指南
一般說明
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
應用
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
特點和優勢
Easily replaceable electrodes, automatic shut off feature
法律資訊
Ohaus is a registered trademark of Ohaus Corp.