Key Documents
LIDF451811-EP
Lid for Eppendorf(R) FA-45-18-11-Kit Rotor
not aerosol-tight, for 5424/5424R, polypropylene
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103913
材料
polypropylene
特點
not aerosol-tight
製造商/商標名
Eppendorf® 022653105 (NA)
Eppendorf® 5424707009 (EU)
技術
centrifugation: suitable
相容性
for use with 5424 R
for use with 5424
法律資訊
Eppendorf is a registered trademark of Eppendorf AG