YY5500053-M
1 of 10
Supelco
1.02408
三(羟甲基)氨基甲烷
Millipore
AC874
NovaSeptum® SURe Bottle Sampling Unit
1.99003
缓冲溶液
1.08382
1.09476
Buffer solution
XX4504700
HP过滤器支架,47 mm,不锈钢
SAFC
1.37092
七水合磷酸氢二钠
Sigma-Aldrich
BI0830
Deblocking Reagent 3% Trichloroacetic Acid in Dichloromethane
1.00030
乙腈
1.02410
碳酸钙
3 in.
SP1
for use with MILLIDISK® 10/40 Cartridge Housings