Key Documents
MXRPCCEI
Millipore
Power cord, Power MIX, all sizes, Chile, Ethiopia, Italy, 250V, 10A
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103801
eCl@ss:
32010600
AC/DC 輸入
250 V, 10 A
品質等級
相容性
for use with MOBIUS® Power MIX - all sizes
法律資訊
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany