87025022
1 of 10
Millipore
87025015
Vantage S2 and A2 Flow Cell Kit V250
96100014
Vantage L Seal/Bed Support Kit
87000003
Vantage S2 and A2 Sanitary Pinch Valves 2-way 6 mm
87018015
Vantage S2 and A2 Flow Cell Kit V180
96100050
Vantage L Tube Kit 250 mm
31138516WB
gasket, u section seal, id 1363mm, epdm peroxyde cured
87000004-M
Vantage S2 and A2 60 90 130 Connector Kit
87025018
Vantage S2 and A2 Chrom Tubes V250
87009015
Vantage S2 and A2 Flow Cell Kit V90
31414305WB
GASKET, U SECTION SEAL, ID 1318MM, EPDM PEROXYDE CURED
Vantage
protein purification: suitable