主要文件
Z291668
Supelco
用于 12×32mm 标准自动进样器样品瓶的螺纹瓶盖
black polypropylene, open-top, 8-425 thread, pkg of 100 ea
别名:
开孔聚丙烯螺纹瓶盖
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122001
NACRES:
NB.11
物料
black polypropylene
Agency
suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010
管理合规性
suitable for FDA C-010.02
特点
autoclavable
closure type black polypropylene hole cap
包装
pkg of 100 ea
参数
135 °C temp. limit
配件
8-425 thread
应用
PFAS testing
一般描述
这些开孔瓶盖均不带隔垫。
法律信息
ASTM is a registered trademark of American Society for Testing and Materials
历史批次信息供参考:
分析证书(COA)