Merck
Z261319
Stainless steel 316 syringe needle, noncoring point
gauge 18, L 6 in.
登录查看公司和协议定价
About This Item
UNSPSC代码:
42142600
eCl@ss:
32070500
NACRES:
NB.03
描述
Micro-Mate Hub with 316 Stainless Steel Tubing
包装
pack of 1 ea
制造商/商品名称
Popper 9860
长度
6 in.
计量器
18
正在寻找类似产品? 访问 产品对比指南
一般描述
Specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.
Micro-Mate Hub with 316 Stainless Steel Tubing
历史批次信息供参考:
分析证书(COA)