主要文件
Z178667
Hamilton® laboratory tubing clamp
fits vessels with rim thickness < 1/8 in., 12-18 gauge tubing size.
登录查看公司和协议定价
About This Item
UNSPSC代码:
41105108
NACRES:
NB.51
制造商/商品名称
Hamilton
一般描述
Snap to rim of any laboratory vessel with rim thickness less than 1/8 in. to secure tubing from 12 to 18 gauge in size.
法律信息
Hamilton is a registered trademark of Hamilton Co.
历史批次信息供参考:
分析证书(COA)