Merck
Z116998
Stainless steel 304 syringe needle
gauge 12, L 12 in.
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122002
NACRES:
NB.03
描述
Standard Hub with 304 Stainless Steel Tubing
包装
pack of 1 ea
制造商/商品名称
Cadence Science
长度
12 in.
计量器
12
正在寻找类似产品? 访问 产品对比指南
一般描述
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
历史批次信息供参考:
分析证书(COA)