Key Documents
CAD9922
Animal feeding needle, sterile, disposable, PTFE
size 17G, L × diam. 2 in. × 2.4 mm , ball
登录查看公司和协议定价
About This Item
分類程式碼代碼:
42142609
NACRES:
NB.03
尺寸
17G
包裝
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
長度 × 直徑
2 in. × 2.4 mm , ball
正在寻找类似产品? 访问 产品对比指南
一般說明
  • PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
  • Plastic Luer hubs and soft silicone tips
  • Non-toxic and no-pyrogenic
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box