主要文件
BR701374
BRAND®微量板密封膜
self-adhesive, suitable for storage, vinyl (with pre-pierced zone)
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122410
NACRES:
NB.35
物料
film (PP/PE-HD)
vinyl (with pre-pierced zone)
无菌性
non-sterile
特点
self-adhesive
包装
pack of 100 ea
制造商/商品名称
BRAND 701374
适用性
suitable for storage
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Due to the increasing number of analysis methods, the number of samples processed is growing steadily. This means that high-throughput analyses are gaining significance. In these automated processes, the films must not only protect the samples, but also comply with the mechanical requirements of automation.
特点和优势
  • Pierceability
  • Adhesive-free zones for contamination-free tasks
  • Tight seal for minimizing evaporation
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)