Key Documents
BR29120
BRAND®移液管冲洗系统,移液管冲洗器
high-density polyethylene, capacity 13 L, L × I.D. 740 mm × 150 mm
别名:
移液管, 移液管冲洗器, 移液管冲洗系统
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41103200
NACRES:
NB.35
材料
high-density polyethylene
製造商/商標名
BRAND 29120
長度 × 內徑
740 mm × 150 mm
容量
13 L
正在寻找类似产品? 访问 产品对比指南
一般說明
PE-HD. Trouble-free and gentle cleaning of pipettes with repetitive siphon rinsing. Water supply connection with hose nozzle outer Ø 12 mm. Soaking jar and pipette basket sold separately. Adjust the water flow to approx. 2 L/min.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG