Key Documents
BR28925
BRAND®铝质移液管盒
Holds 235-305 mm pipettes
别名:
Pipette container, Pipette holder, Storage case
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41123300
NACRES:
NB.13
材料
aluminum
特點
Holds 235-305 mm pipettes
包裝
pack of 1 ea
製造商/商標名
BRAND 28925
正在寻找类似产品? 访问 产品对比指南
一般說明
Pipette box of aluminum, inner silicone coating for storage and sterilisation of pipettes with different length.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG