Key Documents
BR13938
BRAND®备用滴定管延长管,适用于紧凑型滴定管
size 50 mL, SILBERBRAND, amber AR-Glass
别名:
SILBERBRAND, 滴定管, 紧凑型滴定管, 配件
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121808
NACRES:
NB.43
材料
amber AR-Glass
包裝
pack of 1 ea
製造商/商標名
BRAND 13938
長度
610 mm
尺寸
50 mL
正在寻找类似产品? 访问 产品对比指南
一般說明
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 clear glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG