Merck
BR122554
BRAND®实验室瓶子,有刻度和螺口盖
capacity 500 mL
别名:
实验室瓶子, 玻璃, 瓶子
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
材料
borosilicate glass bottle
clear bottle
narrow-mouth bottle
polypropylene cap
round bottle
接合處
Joints threaded neck (GL45)
無菌
non-sterile
特點
autoclavable
graduations
with cap
包裝
pack of 10 ea
製造商/商標名
BRAND 122554
容量
500 mL
直徑× 高度
86 mm × 183 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Threaded to DIN ISO 4796. Studded base for stability and abrasion resistance. Graduated, with marking area. Screw cap and pouring ring PP. Autoclavable (121 °C).
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG