主要文件
BAF133740002
SP Bel-Art® Belpen Marker Pen
blue
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122800
NACRES:
NB.43
物料
blue
制造商/商品名称
Bel-Art F13374-0002
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Felt tip marking pens are perfect for labeling all types of glassware and plastics.
特点和优势
  • Permanent, quick drying ink remains waterproof and smudge proof
  • Will withstand autoclaving when dry
  • Pen with handy pocket clip is 13.3cm (5¹/4") long
法律信息
Bel-Art is a registered trademark of SP Scienceware, a division of SP
历史批次信息供参考:
分析证书(COA)