- Manipulating Cellular Activities Using an Ultrasound-Chemical Hybrid Tool.
Manipulating Cellular Activities Using an Ultrasound-Chemical Hybrid Tool.
ACS synthetic biology (2017-09-26)
Ching-Hsiang Fan, Yao-Shen Huang, Wei-En Huang, Albert Alexander Lee, Sheng-Yang Ho, Yu-Lin Kao, Cuei-Ling Wang, Yen-Ling Lian, Tasuku Ueno, Tsung-Shing Andrew Wang, Chih-Kuang Yeh, Yu-Chun Lin
PMID28945972
ABSTRACT
We developed an ultrasound-chemical hybrid tool to precisely manipulate cellular activities. A focused ultrasound coupled with gas-filled microbubbles was used to rapidly trigger the influx of membrane-impermeable chemical dimerizers into living cells to regulate protein dimerization and location without inducing noticeable toxicity. With this system, we demonstrated the successful modulation of phospholipid metabolism triggered by a short pulse of ultrasound exposure. Our technique offers a powerful and versatile tool for using ultrasound to spatiotemporally manipulate the cellular physiology in living cells.