Merck
  • Use of Bru-Seq and BruChase-Seq for genome-wide assessment of the synthesis and stability of RNA.
Use of Bru-Seq and BruChase-Seq for genome-wide assessment of the synthesis and stability of RNA.
Methods (San Diego, Calif.) (2013-08-27)
Michelle T Paulsen, Artur Veloso, Jayendra Prasad, Karan Bedi, Emily A Ljungman, Brian Magnuson, Thomas E Wilson, Mats Ljungman
ABSTRACT
Gene expression studies commonly examine total cellular RNA, which only provides information about its steady-state pool of RNA. It remains unclear whether differences in the steady-state reflects variable rates of transcription or RNA degradation. To specifically monitor RNA synthesis and degradation genome-wide, we developed Bru-Seq and BruChase-Seq. These assays are based on metabolic pulse-chase labeling of RNA using bromouridine (Bru). In Bru-Seq, recently labeled RNAs are sequenced to reveal spans of nascent transcription in the genome. In BruChase-Seq, cells are chased in uridine for different periods of time following Bru-labeling, allowing for the isolation of RNA populations of specific ages. Here we describe these methodologies in detail and highlight their usefulness in assessing RNA synthesis and stability as well as splicing kinetics with examples of specific genes from different human cell lines.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Pricing and availability is not currently available.