Merck
  • Pharmacokinetics, biodistribution, efficacy and safety of N-octyl-O-sulfate chitosan micelles loaded with paclitaxel.
Pharmacokinetics, biodistribution, efficacy and safety of N-octyl-O-sulfate chitosan micelles loaded with paclitaxel.
Biomaterials (2007-12-21)
Can Zhang, Guowei Qu, Yingji Sun, Xiaoli Wu, Zhong Yao, Qinglong Guo, Qilong Ding, Shengtao Yuan, Zilong Shen, Qineng Ping, Huiping Zhou
ABSTRACT
Paclitaxel (Taxol), PTX) is a promising anti-cancer drug and has been successfully used to treat a wide variety of cancers. Unfortunately, serious clinical side effects are associated with it, which are caused by PTX itself and non-aqueous vehicle containing Cremophor EL. Development of new formulation of PTX with better efficacy and fewer side effects is extremely urgent. In the present study, a N-octyl-O-sulfate chitosan (NOSC) micelle was developed and used as the delivery system for PTX. The pharmacokinetics, biodistribution, efficacy and safety of PTX-loaded NOSC micelles (PTX-M) were evaluated. The results showed that NOSC micelles had high drug loading capacity (69.9%) and entrapment efficiency (97.26%). The plasma AUC of PTX-M was 3.6-fold lower than that of Taxol; but the V(d) and CL of PTX-M were increased by 5.7 and 3.5-fold, respectively. Biodistribution study indicated that most of the PTX were distributed in liver, kidney, spleen, and lung and the longest retention effect was observed in the lung. Drug safety assessment studies including acute toxicity, hemolysis test, intravenous stimulation and injection anaphylaxis revealed that the PTX-M was safe for intravenous injection. Furthermore, the comparable antitumor efficacy of PTX-M and Taxol was observed at the same dose of 10 mg/kg in in vivo antitumor mice models inoculated with sarcoma180, enrich solid carcinoma (EC), hepatoma solidity (Heps), Lewis lung cancer cells and A-549 human lung cancer cells. These results clearly showed that PTX-M had the similar antitumor efficacy as Taxol, but significantly reduced the toxicity and improved the bioavailability of PTX.
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Pricing and availability is not currently available.