Skip to Content
Merck
Thidiazuron PESTANAL®, analytical standard

Thidiazuron

Thidiazuron PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C9H8N4OS
CAS Number:
Molecular Weight:
220.25
EC Number:

Products

Product Category
Formula Weight
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing