
DMT-2′O-Methyl-rG(ib) Phosphoramidite

Synonym(s):
DMT-2′-O-Me-rG(ib) amidite, N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-methyl-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]
Empirical Formula (Hill Notation):
C45H56N7O9P
CAS Number:
Molecular Weight:
869.94
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing