
Corticotropin Releasing Factor Antagonist

Synonym(s):
α-Helical CRF (9-41), α-Helical corticotropin releasing factor fragment (9-41)
Empirical Formula (Hill Notation):
C166H274N46O53S2
CAS Number:
Molecular Weight:
3826.36
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1